missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™
73.94 € - 309.00 €
Chemical Identifiers
CAS | 436799-33-6 |
---|---|
Molecular Formula | C6H3BrF3N |
Molecular Weight (g/mol) | 226 |
MDL Number | MFCD04972700 |
InChI Key | HEDHNDVPKRVQPN-UHFFFAOYSA-N |
Synonym | 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin |
PubChem CID | 11127991 |
IUPAC Name | 3-bromo-5-(trifluoromethyl)pyridine |
SMILES | C1=C(C=NC=C1Br)C(F)(F)F |
Description
Chemical Identifiers
436799-33-6 | |
226 | |
HEDHNDVPKRVQPN-UHFFFAOYSA-N | |
11127991 | |
C1=C(C=NC=C1Br)C(F)(F)F |
C6H3BrF3N | |
MFCD04972700 | |
3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin | |
3-bromo-5-(trifluoromethyl)pyridine |
Specifications
436799-33-6 | |
100.0 | |
White to Yellow | |
85°C | |
96% min. (GC) | |
MFCD04972700 | |
HEDHNDVPKRVQPN-UHFFFAOYSA-N | |
3-bromo-5-(trifluoromethyl)pyridine | |
11127991 | |
97% | |
3-Bromo-5-(trifluoromethyl)pyridine |
96.0 | |
43.5°C to 46.0°C | |
65.0°C to 66.0°C (19.0 mmHg) | |
Authentic | |
C6H3BrF3N | |
3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin | |
C1=C(C=NC=C1Br)C(F)(F)F | |
226 | |
226 | |
Crystalline Powder or Solid |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 3-Bromo-5-(trifluoromethyl)pyridine
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only