missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-6-chlorochromone, 96%, Thermo Scientific™

Product Code. 15484717
Click to view available options
Quantity:
250 mg
2 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73220-38-9
Molecular Formula C9H4BrClO2
Molecular Weight (g/mol) 259.48
MDL Number MFCD02093123
InChI Key ZILSBPAUJJBEFF-UHFFFAOYSA-N
Synonym 3-bromo-6-chloro-4h-chromen-4-one, 3-bromo-6-chlorochromone, 4h-1-benzopyran-4-one,3-bromo-6-chloro, 3-bromo-6-chloro chromone, 3-bromo-6-chloro-4-oxo-4h-1-benzopyran, 3-bromo-6-chloro-4h-1-benzopyran-4-one
PubChem CID 3498945
IUPAC Name 3-bromo-6-chlorochromen-4-one
SMILES ClC1=CC=C2OC=C(Br)C(=O)C2=C1

Specifications

Melting Point 128°C to 130°C
Quantity 250 mg
Formula Weight 259.49
Percent Purity 96%
Chemical Name or Material 3-Bromo-6-chlorochromone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.