missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-7-nitroindole, 97%, Thermo Scientific™

Product Code. 15440929
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 397864-11-8
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
MDL Number MFCD02684155
InChI Key ZJCDMQUXOJHHBJ-UHFFFAOYSA-N
PubChem CID 17750599
IUPAC Name 3-bromo-7-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])NC=C2Br

Specifications

Quantity 250 mg
Synonym 3-bromo-7-nitroindole, 1h-indole, 3-bromo-7-nitro, 1h-indole,3-bromo-7-nitro, 6-methoxy-quinolin-3-ylamine
Formula Weight 241.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-7-nitroindole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.