missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-o-xylene, 99%, Thermo Scientific™

Product Code. 10409020 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
This item is not returnable. View return policy

Product Code. 10409020

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 576-23-8
CAS Min % 98.5
Color Colorless
CAS Max % 100.0
Density 1.3600g/mL
Boiling Point 214°C
Quantity 5 g
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5590 to 1.561
Assay Percent Range 99%
Specific Gravity 1.36
Molecular Formula C8H9Br
Linear Formula (CH3)2C6H3Br
MDL Number MFCD00000069
Beilstein 05, 365
Synonym 3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
InChI Key WLPXNBYWDDYJTN-UHFFFAOYSA-N
SMILES CC1=CC=CC(Br)=C1C
IUPAC Name 1-bromo-2,3-dimethylbenzene
Molecular Weight (g/mol) 185.06
PubChem CID 68472
Formula Weight 185.06
Percent Purity 99%
Chemical Name or Material 3-Bromo-o-xylene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-o-xylene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.