missing translation for 'onlineSavingsMsg'
Learn More
3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals Product Code.: 11495557

3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals

Product Code. 11495557
1 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11495557

Brand: Thermo Scientific Alfa Aesar B23543.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuran was a synthesis reagent used in the preparation of 2-substituted 3-furfurals. It was also used in the synthesis of 5,6-dehydronorcantharidins. It is one of the volatile metabolites discussed in studies for detecting and discriminating diseases of onion.

Solubility
Soluble in chloroform, ethyl acetate. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from heat, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22037-28-1
Molecular Formula C4H3BrO
Molecular Weight (g/mol) 146.97
MDL Number MFCD00005347
InChI Key LXWLEQZDXOQZGW-UHFFFAOYSA-N
Synonym furan, 3-bromo, .beta.-bromofuran, beta-bromofuran, 3-furyl bromide, 3-bromo furan, 3-bromo-furan, zlchem 308, pubchem8830, 3-bromofuran, acmc-209fqzShow MoreShow Less
PubChem CID 89164
IUPAC Name 3-bromofuran
SMILES BrC1=COC=C1

Specifications

Density 1.635
Boiling Point 102°C to 104°C
Flash Point 3°C (37°F)
Refractive Index 1.496
Quantity 1 g
UN Number UN1993
Beilstein 105337
Solubility Information Soluble in chloroform,ethyl acetate. Insoluble in water.
Formula Weight 146.98
Percent Purity 97%
Chemical Name or Material 3-Bromofuran, stabilized with 0.5% calcium carbonate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberD25S032 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23543.06,B23543.03,B23543.14
Lot Number10181871 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23543.06,B23543.03,B23543.14
Lot Number10180659 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23543.06,B23543.03,B23543.14
Lot Number10153307 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23543.06,B23543.03,B23543.14
Lot Number10105557 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB23543.06,B23543.03,B23543.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromofuran, 97%, stab. with 0.5% calcium carbonate, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.