missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromoimidazo[1,2-a]pyridine, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber glass bottle |
| Quantity | 5 g |
| Synonym | 3-bromoimidazo 1,2-a pyridine, 3-bromo-imidazo 1,2-a pyridine, 3-bromopyrimidazole, imidazo 1,2-a pyridine, 3-bromo, 3-bromoimidazolo 1,2-a pyridine, 3-bromoh-imidazo 1,2-a pyridine, pubchem17183, acmc-209kfj, buttpark 96\50-11, imidazo 1,2-a pyridine,3-bromo |
| Formula Weight | 197.03 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-Bromoimidazo[1, 2-a]pyridine |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction