missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-5-chlorobenzo[b]thiophene, 95+%, Thermo Scientific™

Product Code. 10205844
Click to view available options
Quantity:
1 g
5 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 1198-51-2
Molecular Formula C9H6BrClS
Molecular Weight (g/mol) 261.56
MDL Number MFCD00173769
InChI Key FKQSFVITUNJLCY-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-chlorobenzo b thiophene, 3-bromomethyl-5-chloro-1-benzothiophene, 3-bromomethyl-5-chloro benzo b thiophene, 3-bromomethyl-5-chlorobenzothiophene, 5-chloro-3-bromomethylbenzothiophene, 5-chloro-3-bromomethyl-benzo b-thiophene, benzo b thiophene, 3-bromomethyl-5-chloro, pubchem13553
PubChem CID 2779857
IUPAC Name 3-(bromomethyl)-5-chloro-1-benzothiophene
SMILES ClC1=CC=C2SC=C(CBr)C2=C1

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 261.57
Percent Purity 95%
Chemical Name or Material 3-(Bromomethyl)-5-chlorobenzo[b]thiophene
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.