missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropylamine hydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 10274560
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5003-71-4
Molecular Formula C3H8BrN·HBr
Molecular Weight (g/mol) 218.93
MDL Number MFCD00012912
InChI Key PQIYSSSTRHVOBW-UHFFFAOYSA-N
Synonym 3-bromopropylamine hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-aminopropyl bromide hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-bromopropylammonium bromide, 3-bromopropanamine hydrobromide, 1-amino-3-bromopropane hydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-brominepropylaminehydrobromide, 3-bromopropylamine, hydrobromide
PubChem CID 78701
IUPAC Name 3-bromopropan-1-amine;hydrobromide
SMILES C(CN)CBr.Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 171.0°C to 175.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula Br(CH2)3NH2·HBr
Quantity 100 g
Beilstein 04, 149
Solubility Information Solubility in water: soluble
Formula Weight 218.93
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 3-Bromopropylamine hydrobromide, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromopropylamine hydrobromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'