missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromothiophene-2-carboxaldehyde diethyl acetal, 97%, Thermo Scientific™
59.60 € - 206.00 €
Chemische Identifikatoren
CAS | 34042-95-0 |
---|---|
Molecular Formula | C9H13BrO2S |
Molecular Weight (g/mol) | 265.165 |
MDL Number | MFCD03426030 |
InChI Key | QNTDBERHEOAJAA-UHFFFAOYSA-N |
Synonym | 3-bromothiophene-2-carboxaldehyde diethyl acetal, 3-bromo-2-diethoxymethyl thiophene, acmc-1aiew, 3-bromo 2-thienyl diethoxymethane, 3-bromothiophene-2-carbaldehyde diethyl acetal, 3-bromothiophene-2carboxaldehyde diethyl acetal, 3-bromo-2-thiophenecarboxaldehyde diethyl acetal |
PubChem CID | 7022388 |
IUPAC Name | 3-bromo-2-(diethoxymethyl)thiophene |
SMILES | CCOC(C1=C(C=CS1)Br)OCC |
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
---|---|---|---|---|---|---|---|---|---|
Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
11358686
|
Thermo Scientific Alfa Aesar
L19397.03 |
1 g |
59.60 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11368686
|
Thermo Scientific Alfa Aesar
L19397.06 |
5 g |
206.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemische Identifikatoren
34042-95-0 | |
265.165 | |
QNTDBERHEOAJAA-UHFFFAOYSA-N | |
7022388 | |
CCOC(C1=C(C=CS1)Br)OCC |
C9H13BrO2S | |
MFCD03426030 | |
3-bromothiophene-2-carboxaldehyde diethyl acetal, 3-bromo-2-diethoxymethyl thiophene, acmc-1aiew, 3-bromo 2-thienyl diethoxymethane, 3-bromothiophene-2-carbaldehyde diethyl acetal, 3-bromothiophene-2carboxaldehyde diethyl acetal, 3-bromo-2-thiophenecarboxaldehyde diethyl acetal | |
3-bromo-2-(diethoxymethyl)thiophene |
Spezifikation
34042-95-0 | |
C9H13BrO2S | |
MFCD03426030 | |
3-bromothiophene-2-carboxaldehyde diethyl acetal, 3-bromo-2-diethoxymethyl thiophene, acmc-1aiew, 3-bromo 2-thienyl diethoxymethane, 3-bromothiophene-2-carbaldehyde diethyl acetal, 3-bromothiophene-2carboxaldehyde diethyl acetal, 3-bromo-2-thiophenecarboxaldehyde diethyl acetal | |
CCOC(C1=C(C=CS1)Br)OCC | |
265.165 | |
265.17 | |
3-Bromothiophene-2-carboxaldehyde diethyl acetal |
70°C (0.9 mmHg) | |
1.524 | |
1284657 | |
QNTDBERHEOAJAA-UHFFFAOYSA-N | |
3-bromo-2-(diethoxymethyl)thiophene | |
7022388 | |
97% |
Sicherheit und Handhabung
GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362
H315-H319
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures