missing translation for 'onlineSavingsMsg'
Learn More
3-Bromothiophene, 97%, Thermo Scientific Chemicals Product Code.: 11407906

3-Bromothiophene, 97%, Thermo Scientific Chemicals

Product Code. 11407906
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy

Product Code. 11407906

Brand: Thermo Scientific Alfa Aesar A14022.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, μ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene is used in the preparation of derivatives such as thienylenic alpha, µ-diformyl-alpha-oligothiophenes and 3-lithiothiophene. It is also used in the synthesis of 3-lithiothiophene by reacting with n-butyllithium.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong reducing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 872-31-1
Molecular Formula C4H3BrS
Molecular Weight (g/mol) 163.03
MDL Number MFCD00005464
InChI Key XCMISAPCWHTVNG-UHFFFAOYSA-N
Synonym 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306
PubChem CID 13383
IUPAC Name 3-bromothiophene
SMILES BrC1=CSC=C1

Specifications

Density 1.722
Boiling Point 157°C to 158°C
Flash Point 63°C (145°F)
Odor Pungent
Refractive Index 1.591
Quantity 250 g
UN Number UN2810
Beilstein 105338
Solubility Information Immiscible with water.
Formula Weight 163.04
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10214061 Certificate TypeCertificate of Analysis Date16/01/2024 Catalog NumberA14022.30,A14022.09,A14022.18
Lot Number10243704 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA14022.30,A14022.09,A14022.18
Lot Number10206856 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14022.30,A14022.09,A14022.18
Lot Number10155360 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14022.30,A14022.09,A14022.18
Lot Number10138969 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14022.30,A14022.09,A14022.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromothiophene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.