Learn More
3-Bromothiophene, 97%, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 872-31-1 |
---|---|
Molecular Formula | C4H3BrS |
Molecular Weight (g/mol) | 163.03 |
MDL Number | MFCD00005464 |
InChI Key | XCMISAPCWHTVNG-UHFFFAOYSA-N |
Synonym | 3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306 |
PubChem CID | 13383 |
IUPAC Name | 3-bromothiophene |
SMILES | BrC1=CSC=C1 |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
872-31-1 | |
163.03 | |
XCMISAPCWHTVNG-UHFFFAOYSA-N | |
13383 | |
BrC1=CSC=C1 |
C4H3BrS | |
MFCD00005464 | |
3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306 | |
3-bromothiophene |
Specifications
872-31-1 | |
1.7400g/mL | |
56°C | |
96% min. (GC) | |
1.5905 to 1.5925 | |
1.74 | |
Solubility in water: immiscible. Other solubilities: soluble in acetone,benzene,slightly soluble in chloroform | |
BrC1=CSC=C1 | |
163.03 | |
163.03 | |
Liquid |
Colorless to Yellow | |
158.0°C | |
Authentic | |
C4H3BrS | |
MFCD00005464 | |
3-thienyl bromide, thiophene, 3-bromo, 3-bromo thiophene, beta-bromothiophene, .beta.-bromothiophene, 3-bromo-thiophene, bromothiophene 3-, 3-bromothiphene, 3-bromthiophene, pubchem5306 | |
XCMISAPCWHTVNG-UHFFFAOYSA-N | |
3-bromothiophene | |
13383 | |
97% | |
3-Bromothiophene |
Safety and Handling
GHS H Statement
Fatal in contact with skin.
Causes skin irritation.
Fatal if inhaled.
Toxic to aquatic life with long lasting effects.
Causes serious eye irritation.
Toxic if swallowed.
May cause respiratory irr
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Wash face,hands and any exposed skin thoroughly
GHS Signal Word: Danger
missing translation for 'einecsNumber' : 212-821-3
Recommended Storage : Product may darken on storage
- 3-Bromothiophene
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only