Learn More
3-Buten-1-amine, tech. 90%, Thermo Scientific Chemicals
119.00 € - 460.00 €
Chemical Identifiers
CAS | 2524-49-4 |
---|---|
Molecular Formula | C4H9N |
Molecular Weight (g/mol) | 71.12 |
MDL Number | MFCD03425859 |
InChI Key | ASVKKRLMJCWVQF-UHFFFAOYSA-N |
Synonym | 3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene |
PubChem CID | 443732 |
ChEBI | CHEBI:31108 |
IUPAC Name | but-3-en-1-amine |
SMILES | NCCC=C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11378966
|
Thermo Scientific Alfa Aesar
L20157.03 |
1 g |
119.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11388966
|
Thermo Scientific Alfa Aesar
L20157.06 |
5 g |
460.00 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimer’s disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Buten-1-amine is an aliphatic terminal amine used for organic synthesis and in proteomics research. Its hydrochloride salt is used to improve the detection of neurodegenerative alzheimer’s disease in ELISA titrations by reducing the background due to non-specific associations of molecules and proteins.
Solubility
Miscible with water.
Notes
Incompatible with strong acids, strong oxidizing agents and strong bases.
Chemical Identifiers
2524-49-4 | |
71.12 | |
ASVKKRLMJCWVQF-UHFFFAOYSA-N | |
443732 | |
but-3-en-1-amine |
C4H9N | |
MFCD03425859 | |
3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene | |
CHEBI:31108 | |
NCCC=C |
Specifications
2524-49-4 | |
74°C to 77°C | |
Amine-like | |
1.424 | |
UN2733 | |
Air Sensitive | |
Miscible with water. | |
NCCC=C | |
71.12 | |
CHEBI:31108 | |
97% |
0.777 | |
−5°C (23°F) | |
C4H9N | |
MFCD03425859 | |
1735803 | |
3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene | |
ASVKKRLMJCWVQF-UHFFFAOYSA-N | |
but-3-en-1-amine | |
443732 | |
71.12 | |
3-Buten-1-amine |
Safety and Handling
GHS H Statement
H225-H314-H318
Highly flammable liquid and vapor.
Causes severe skin burns and eye damage.
Causes serious eye damage.
P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c
H225-H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: AMINES, FLAMMABLE, CORROSIVE, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.