missing translation for 'onlineSavingsMsg'
Learn More

3-Butenylamine, 97%, Thermo Scientific Chemicals

117.59 € - 403.01 €

Chemical Identifiers

CAS 2524-49-4
Molecular Formula C4H9N
Molecular Weight (g/mol) 71.12
MDL Number MFCD03425859
InChI Key ASVKKRLMJCWVQF-UHFFFAOYSA-N
Synonym 3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene
PubChem CID 443732
ChEBI CHEBI:31108
IUPAC Name but-3-en-1-amine
SMILES NCCC=C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15204428
View Documents
Thermo Scientific Acros
455050010
1 g
117.59 €
1g
Please to purchase this item. Need a web account? Register with us today!
15214428
View Documents
Thermo Scientific Acros
455050050
5 g
403.01 €
5g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2524-49-4
71.12
ASVKKRLMJCWVQF-UHFFFAOYSA-N
443732
but-3-en-1-amine
C4H9N
MFCD03425859
3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene
CHEBI:31108
NCCC=C

Specifications

2524-49-4
−5°C
96% min. (GC)
C4H9N
MFCD03425859
Solubility in water: soluble.
NCCC=C
71.12
CHEBI:31108
97%
83.0°C
Authentic
Glass Bottle
H2C=CH(CH2)2NH2
3-buten-1-amine, homoallylamine, 3-butenylamine, but-3-enylamine, acmc-1b0ze, 4-amino-1-butene, 3-buten-1-amine 9ci, n-but-3-enylamine, 3-butene-1-amine, 1-aminobut-3-ene
ASVKKRLMJCWVQF-UHFFFAOYSA-N
but-3-en-1-amine
443732
71.12
3-Butenylamine
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Highly flammable liquid and vapour.
Causes severe skin burns and eye damage.

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN (or ha

GHS Signal Word: Danger

Product Identifier
  • 3-Butenylamine

Signal Word
  • Danger

Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B

Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.