Learn More
STEMCELL Technologies 3 cc Syringes
For accurate dispensing of methylcellulose-based medium
Brand: STEMCELL Technologies 28240
This item is not returnable.
View return policy
Description
- Ideal for accurate dispensing of methylcellulose-based medium (e.g. MethoCult™), when used with Blunt-End Needles (Catalog #28110) purchased from STEMCELL Technologies to ensure compatibility and accuracy
Specifications
Colony Assay | |
100 Syringes/Bag |