missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10357033
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 175277-52-8
Molecular Formula C7H4Cl2F3N
Molecular Weight (g/mol) 230.011
MDL Number MFCD00173852
InChI Key NUSXEDNPFNWFDQ-UHFFFAOYSA-N
PubChem CID 22058750
IUPAC Name 3-chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC(=C1Cl)CCl)C(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Synonym 3-chloro-2-chloromethyl-5-trifluoromethyl pyridine, 3-chloranyl-2-chloromethyl-5-trifluoromethyl pyridine, pyridine,3-chloro-2-chloromethyl-5-trifluoromethyl, pyridine, 3-chloro-2-chloromethyl-5-trifluoromethyl
Formula Weight 230.02
Percent Purity 95%
Chemical Name or Material 3-Chloro-2-(chloromethyl)-5-(trifluoromethyl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.