missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-2-fluorophenol, 97%, Thermo Scientific™

Product Code. 11372699
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Chloro-2-fluorophenol are employed in the preparation of resins, dyes, explosives, lubricants, and plastics. Also they are are endowed with antioxidant property and are used as antioxidants in many formulations such as pharmaceuticals, cosmetics, electrical transformer oil, solvents, and organic reactions.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2613-22-1
Molecular Formula C6H4ClFO
Molecular Weight (g/mol) 146.545
MDL Number MFCD01631572
InChI Key PCHPYNHSMSAJEU-UHFFFAOYSA-N
Synonym 3-chloro-2-fluoro-phenol, phenol, 3-chloro-2-fluoro, acmc-20agxt, pubchem2855, 3-chloro-2-fluorophe, 2-fluoro-3-chloro-phenol, intermediates-zcf02697, phenol,3-chloro-2-fluoro, ksc557c2h, 3-chloro-2-fluorophenol
PubChem CID 2773712
IUPAC Name 3-chloro-2-fluorophenol
SMILES C1=CC(=C(C(=C1)Cl)F)O

Specifications

Melting Point 34°C to 38°C
Boiling Point 190°C
Odor Phenol-like
Quantity 5 g
UN Number UN3261
Solubility Information Insoluble in water.
Formula Weight 146.55
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorophenol
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'