missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-(3-pyridyl)-1,2,5-thiadiazole, 95%, Thermo Scientific Chemicals

Product Code. 15474857
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 131986-28-2
Molecular Formula C7H4ClN3S
Molecular Weight (g/mol) 197.64
MDL Number MFCD03066187
InChI Key CMPNWGQBNRHIQZ-UHFFFAOYSA-N
Synonym 3-chloro-4-pyridin-3-yl-1,2,5-thiadiazole, 3-chloro-4-3-pyridyl-1,2,5-thiadiazole, 3-4-chloro-1,2,5-thiadiazol-3-yl pyridine, pyridine, 3-4-chloro-1,2,5-thiadiazol-3-yl, 3-4-chloro-1,2,5 thiadiazol-3-yl-pyridine, 3-3-chloro-1,2,5-thiadiazol-4yl pyridine, 3-3-chloro-1,2,5-thiadiazol-4-yl pyridine, acmc-1bz6y, ksc497a5t, 3-chloro-4-pyrid-3-yl-1,2,5-thiadiazole
PubChem CID 10488164
IUPAC Name 3-chloro-4-pyridin-3-yl-1,2,5-thiadiazole
SMILES ClC1=NSN=C1C1=CN=CC=C1

Specifications

Melting Point 56°C
Quantity 250 mg
Formula Weight 197.65
Percent Purity 95%
Chemical Name or Material 3-Chloro-4-(3-pyridyl)-1,2,5-thiadiazole

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'