missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-4-hydroxy-5-methoxybenzyl alcohol, 98%, Thermo Scientific™

19.70 € - 209.00 €

Chemical Identifiers

CAS 20624-92-4
Molecular Formula C8H9ClO3
Molecular Weight (g/mol) 188.61
MDL Number MFCD00016870
InChI Key VQEZBFOBPUKDGM-UHFFFAOYSA-N
Synonym 5-chlorovanillyl alcohol, 2-chloro-4-hydroxymethyl-6-methoxyphenol, 3-chloro-4-hydroxy-5-methoxybenzyl alcohol, 3-chloro-4-hydroxy-5-methoxybenzenemethanol, acmc-20am3y, 6-chloro-4-hydroxymethyl-2-methoxyphenol, 5-chloro-4-hydroxy-3-methoxybenzenemethanol, benzenemethanol,3-chloro-4-hydroxy-5-methoxy
PubChem CID 4181655
IUPAC Name 2-chloro-4-(hydroxymethyl)-6-methoxyphenol
SMILES COC1=C(O)C(Cl)=CC(CO)=C1
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11496137
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B25108.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11406147
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B25108.06
5 g
58.00 €
5g
Please to purchase this item. Need a web account? Register with us today!
11416147
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B25108.14
25 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

20624-92-4
188.61
VQEZBFOBPUKDGM-UHFFFAOYSA-N
4181655
COC1=C(O)C(Cl)=CC(CO)=C1
C8H9ClO3
MFCD00016870
5-chlorovanillyl alcohol, 2-chloro-4-hydroxymethyl-6-methoxyphenol, 3-chloro-4-hydroxy-5-methoxybenzyl alcohol, 3-chloro-4-hydroxy-5-methoxybenzenemethanol, acmc-20am3y, 6-chloro-4-hydroxymethyl-2-methoxyphenol, 5-chloro-4-hydroxy-3-methoxybenzenemethanol, benzenemethanol,3-chloro-4-hydroxy-5-methoxy
2-chloro-4-(hydroxymethyl)-6-methoxyphenol

Specifications

20624-92-4
C8H9ClO3
5-chlorovanillyl alcohol, 2-chloro-4-hydroxymethyl-6-methoxyphenol, 3-chloro-4-hydroxy-5-methoxybenzyl alcohol, 3-chloro-4-hydroxy-5-methoxybenzenemethanol, acmc-20am3y, 6-chloro-4-hydroxymethyl-2-methoxyphenol, 5-chloro-4-hydroxy-3-methoxybenzenemethanol, benzenemethanol,3-chloro-4-hydroxy-5-methoxy
COC1=C(O)C(Cl)=CC(CO)=C1
188.61
188.62
3-Chloro-4-hydroxy-5-methoxybenzyl alcohol
115°C to 118°C
MFCD00016870
VQEZBFOBPUKDGM-UHFFFAOYSA-N
2-chloro-4-(hydroxymethyl)-6-methoxyphenol
4181655
98%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications