missing translation for 'onlineSavingsMsg'
Learn More

3'-Chloro-4'-(trifluoromethoxy)acetophenone, 98%, Thermo Scientific™

61.00 € - 790.00 €

Chemical Identifiers

CAS 129604-27-9
Molecular Formula C9H6ClF3O2
Molecular Weight (g/mol) 238.59
MDL Number MFCD12545820
InChI Key BUSQQCHYIYPNTO-UHFFFAOYSA-N
Synonym 1-3-chloro-4-trifluoromethoxy phenyl ethanone, 3-chloro-4-trifluoromethoxy acetophenone, 3'-chloro-4'-trifluoromethoxyacetophenone, 3'-chloro-4'-trifluoromethoxy acetophenone, intermediates-zcf02001, ethanone, 1-3-chloro-4-trifluoromethoxy phenyl, 1-3-chloro-4-trifluoromethoxy phenyl ethan-1-one
PubChem CID 14793273
IUPAC Name 1-[3-chloro-4-(trifluoromethoxy)phenyl]ethanone
SMILES CC(=O)C1=CC(=C(C=C1)OC(F)(F)F)Cl
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15426766
View Documents
Thermo Scientific Alfa Aesar
H61768.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15458338
View Documents
Thermo Scientific Alfa Aesar
H61768.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15468338
View Documents
Thermo Scientific Alfa Aesar
H61768.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3′-Chloro-4′-(trifluoromethoxy)acetophenone is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications

Chemical Identifiers

129604-27-9
238.59
BUSQQCHYIYPNTO-UHFFFAOYSA-N
14793273
CC(=O)C1=CC(=C(C=C1)OC(F)(F)F)Cl
C9H6ClF3O2
MFCD12545820
1-3-chloro-4-trifluoromethoxy phenyl ethanone, 3-chloro-4-trifluoromethoxy acetophenone, 3'-chloro-4'-trifluoromethoxyacetophenone, 3'-chloro-4'-trifluoromethoxy acetophenone, intermediates-zcf02001, ethanone, 1-3-chloro-4-trifluoromethoxy phenyl, 1-3-chloro-4-trifluoromethoxy phenyl ethan-1-one
1-[3-chloro-4-(trifluoromethoxy)phenyl]ethanone

Specifications

129604-27-9
C9H6ClF3O2
1-3-chloro-4-trifluoromethoxy phenyl ethanone, 3-chloro-4-trifluoromethoxy acetophenone, 3'-chloro-4'-trifluoromethoxyacetophenone, 3'-chloro-4'-trifluoromethoxy acetophenone, intermediates-zcf02001, ethanone, 1-3-chloro-4-trifluoromethoxy phenyl, 1-3-chloro-4-trifluoromethoxy phenyl ethan-1-one
BUSQQCHYIYPNTO-UHFFFAOYSA-N
1-[3-chloro-4-(trifluoromethoxy)phenyl]ethanone
14793273
98%
3'-Chloro-4'-(trifluoromethoxy)acetophenone
Colorless
MFCD12545820
Slightly soluble in water.
CC(=O)C1=CC(=C(C=C1)OC(F)(F)F)Cl
238.59
238.59
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications