missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(pentafluorothio)benzoic acid, 97%, Thermo Scientific™

Product Code. 15486757
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1448317-67-6
Molecular Formula C7H4ClF5O2S
Molecular Weight (g/mol) 282.609
MDL Number MFCD16652415
InChI Key QEUCPMZEYUIRBW-UHFFFAOYSA-N
Synonym 3-chloro-5-pentafluorosulfur benzoic acid, 3-chloro-5-pentafluoro-??-sulfanyl benzoic acid, 3-chloro-5-pentafluorothio benzoic acid, 3-chloro-5-pentafluoro-lambda6-sulphanyl benzoic acid
PubChem CID 66523456
IUPAC Name 3-chloro-5-(pentafluoro-$l^{6}-sulfanyl)benzoic acid
SMILES C1=C(C=C(C=C1S(F)(F)(F)(F)F)Cl)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 282.61
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(pentafluorothio)benzoic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'