missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-5-(trifluoromethyl)pyridine, ≥97%, Thermo Scientific™

Product Code. 10471874
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 85148-26-1
Molecular Formula C6H3ClF3N
Molecular Weight (g/mol) 181.54
MDL Number MFCD00042227
InChI Key OMRCXTBFBBWTDL-UHFFFAOYSA-N
Synonym 3-chloro-5-trifluoromethyl pyridine, 3-trifluoromethyl-5-chloropyridine, pyridine, 3-chloro-5-trifluoromethyl, pubchem3552, acmc-209zee, ksc495c1f, 3-chloro-5-trifluoromethyl-pyridine, 3-chloro-5-trifluoromethyl pyridine-carbonitrile
PubChem CID 2736698
IUPAC Name 3-chloro-5-(trifluoromethyl)pyridine
SMILES FC(F)(F)C1=CC(Cl)=CN=C1

Specifications

CAS Min % 97.0
Assay Percent Range 97+%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 181.54
Percent Purity 97%
Chemical Name or Material 3-Chloro-5-(trifluoromethyl)pyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'