missing translation for 'onlineSavingsMsg'
Learn More

3-Chloro-N-methylaniline, 97%, Thermo Scientific Chemicals

Product Code. 11499603
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7006-52-2
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00052014
InChI Key WFGYSQDPURFIFL-UHFFFAOYSA-N
Synonym n-methyl-3-chloroaniline, 3-chloro-n-methylbenzenamine, benzenamine, 3-chloro-n-methyl, 3-chloro-phenyl-methyl-amine, n-methyl-m-chloroaniline, n1-methyl-3-chloroaniline, pubchem9202, acmc-209ocl, 3-chloro-1-methylaniline, 3-chloro-n-methyl aniline
PubChem CID 138900
IUPAC Name 3-chloro-N-methylaniline
SMILES CNC1=CC=CC(Cl)=C1

Specifications

Density 1.163
Boiling Point 243°C to 244°C
Flash Point >110°C (230°F)
Refractive Index 1.583
Quantity 25 g
UN Number UN2810
Beilstein 2802319
Formula Weight 141.6
Percent Purity 97%
Chemical Name or Material 3-Chloro-N-methylaniline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.