missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzamide, 98+%, Thermo Scientific Chemicals

Product Code. 11460213
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 618-48-4
Molecular Formula C7H6ClNO
Molecular Weight (g/mol) 155.58
MDL Number MFCD00014799
InChI Key MJTGQALMWUUPQM-UHFFFAOYSA-N
Synonym m-chlorobenzamide, benzamide, 3-chloro, 3-chloro-benzamide, benzamide, m-chloro-8ci, benzamide, m-chloro, 5-chlorobenzamide, pubchem3618, acmc-1b3cu, wce
PubChem CID 69254
ChEBI CHEBI:10587
IUPAC Name 3-chlorobenzamide
SMILES NC(=O)C1=CC=CC(Cl)=C1

Specifications

Melting Point 133°C to 136°C
Quantity 10 g
Beilstein 1859940
Formula Weight 155.58
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzamide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'