missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorobenzophenone, 98+%, Thermo Scientific Chemicals

Product Code. 11496276
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1016-78-0
Molecular Formula C13H9ClO
Molecular Weight (g/mol) 216.664
MDL Number MFCD00009816
InChI Key CPLWKNRPZVNELG-UHFFFAOYSA-N
Synonym 3-chlorobenzophenone, 3-chlorophenyl phenyl methanone, benzophenone, 3-chloro, methanone, 3-chlorophenyl phenyl, m-chlorobenzophenone, 3-chlorophenyl-phenylmethanone, methanone, 3-chlorophenyl pheny, 3-chlorophenyl phenyl ketone, 5-chlorobenzophenone, pubchem24414
PubChem CID 66098
IUPAC Name (3-chlorophenyl)-phenylmethanone
SMILES C1=CC=C(C=C1)C(=O)C2=CC(=CC=C2)Cl

Specifications

Melting Point 81°C to 85°C
Quantity 25 g
Beilstein 1869822
Formula Weight 216.67
Percent Purity ≥98%
Chemical Name or Material 3-Chlorobenzophenone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.