missing translation for 'onlineSavingsMsg'
Learn More

3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid, 95%, Thermo Scientific™

Product Code. 15448345
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100001-79-4
Molecular Formula C8H5ClN2O2
Molecular Weight (g/mol) 196.59
MDL Number MFCD09836169
InChI Key JBQOFBOMIUELMB-UHFFFAOYSA-N
Synonym 3-chloroimidazo 1,2-a pyridine-2-carboxylic acid, 3-chloro-4-hydroimidazo 1,2-a pyridine-2-carboxylic acid
PubChem CID 26369186
IUPAC Name 3-chloroimidazo[1,2-a]pyridine-2-carboxylic acid
SMILES C1=CC2=NC(=C(N2C=C1)Cl)C(=O)O

Specifications

Melting Point 203°C to 205°C
Quantity 250 mg
Formula Weight 196.59
Percent Purity 95%
Chemical Name or Material 3-Chloroimidazo[1,2-a]pyridine-2-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'