missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)-5-(2-thienyl)-1,2,4-oxadiazole, ≥95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
Specifications
Specifications
| CAS Min % | 95.0 |
| Assay Percent Range | 95% |
| Quantity | 1 g |
| Formula Weight | 200.65 |
| Percent Purity | 95% |
| Chemical Name or Material | 3-(Chloromethyl)-5-(2-thienyl)-1, 2, 4-oxadiazole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction