missing translation for 'onlineSavingsMsg'
Learn More

3-(Chloromethyl)-5-methylisoxazole, 97%, Thermo Scientific™

Product Code. 10213192
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 35166-37-1
Molecular Formula C5H6ClNO
Molecular Weight (g/mol) 131.56
MDL Number MFCD00221166
InChI Key FEXTXBAFBURKGS-UHFFFAOYSA-N
Synonym 3-chloromethyl-5-methylisoxazole, 3-chloromethyl-5-methyl-1,2-oxazole, 3-chloromethyl-5-methyl-isoxazole, pubchem8710, zlchem 1024, acmc-209idp, 3-chloromethyl-5-methyisoxazole, 3-choromethyl-5-methylisoxazole, 5-methyl-3-chloromethylisoxazole
PubChem CID 2735763
IUPAC Name 3-(chloromethyl)-5-methyl-1,2-oxazole
SMILES CC1=CC(CCl)=NO1

Specifications

Assay Percent Range 97%
Quantity 250 mg
Formula Weight 131.56
Percent Purity 97%
Chemical Name or Material 3-(Chloromethyl)-5-methylisoxazole
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.