missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Chloromethyl)-5-phenylisoxazole, ≥97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
Synonym | 3-chloromethyl-5-phenylisoxazole, 3-chloromethyl-5-phenyl-1,2-oxazole, isoxazole, 3-chloromethyl-5-phenyl, 5-phenyl-3-chloromethyl isoxazole, isoxazole,3-chloromethyl-5-phenyl |
Formula Weight | 193.63 |
Percent Purity | 97% |
Chemical Name or Material | 3-(Chloromethyl)-5-phenylisoxazole |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction