Learn More
(3-Chloropropyl)trimethoxysilane, 97+%, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
29.25 € - 292.00 €
Chemical Identifiers
CAS | 2530-87-2 |
---|---|
Molecular Formula | C6H15ClO3Si |
Molecular Weight (g/mol) | 198.718 |
MDL Number | MFCD00000997 |
InChI Key | OXYZDRAJMHGSMW-UHFFFAOYSA-N |
Synonym | 3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan |
PubChem CID | 62449 |
IUPAC Name | 3-chloropropyl(trimethoxy)silane |
SMILES | CO[Si](CCCCl)(OC)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11354568
|
Thermo Scientific Alfa Aesar
042413.AD |
50 mL |
29.25 €
50mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11334568
|
Thermo Scientific Alfa Aesar
042413.AK |
250 mL |
91.20 €
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11344568
|
Thermo Scientific Alfa Aesar
042413.K2 |
1 L |
292.00 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
(3-Chloropropyl)trimethoxysilane is used as adhesives and sealant chemicals. It is also used in the preparation of 3-iodopropyltrimethoxysilane by reacting with sodium iodide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under Argon
Chemical Identifiers
2530-87-2 | |
198.718 | |
OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
62449 | |
CO[Si](CCCCl)(OC)OC |
C6H15ClO3Si | |
MFCD00000997 | |
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
3-chloropropyl(trimethoxy)silane |
Specifications
195°C to 197°C | |
2530-87-2 | |
C6H15ClO3Si | |
969627 | |
3-chloropropyl trimethoxysilane, silane, 3-chloropropyl trimethoxy, 3-chloropropyltrimethyoxysilane, sila-ace s 620, cps-m, silane 3-chloropropyl tris methoxy, 3-trimethoxysilyl propyl chloride, unii-t21bnl1s7f, cptmo, 3-chloropropyl trimethoxysilan | |
OXYZDRAJMHGSMW-UHFFFAOYSA-N | |
3-chloropropyl(trimethoxy)silane | |
62449 | |
≥97% | |
ChemSeal™ bottles | |
Moisture sensitive | |
(3-Chloropropyl)trimethoxysilane |
-50°C | |
Liquid | |
MFCD00000997 | |
78°C (172°F) | |
Miscible with organic solvents. | |
CO[Si](CCCCl)(OC)OC | |
198.718 | |
198.72 | |
Recognizable | |
1.42 | |
1.077 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
P210-P280-P370+P378q-P501c
H227
missing translation for 'einecsNumber' : 219-787-9
missing translation for 'rtecsNumber' : VV2680000
missing translation for 'tsca' : Yes
missing translation for 'storageNote' :
- Packaged under Argon
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.