Learn More
3-Chloropropylamine hydrochloride, 98%, Thermo Scientific Chemicals
39.40 € - 431.00 €
Chemical Identifiers
CAS | 6276-54-6 |
---|---|
Molecular Formula | C3H9Cl2N |
Molecular Weight (g/mol) | 130.012 |
MDL Number | MFCD00012913 |
InChI Key | IHPRVZKJZGXTBQ-UHFFFAOYSA-N |
Synonym | 3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b |
PubChem CID | 11469095 |
IUPAC Name | 3-chloropropan-1-amine;hydrochloride |
SMILES | C(CN)CCl.Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11458396
|
Thermo Scientific Alfa Aesar
A15415.18 |
50 g |
39.40 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11468396
|
Thermo Scientific Alfa Aesar
A15415.30 |
250 g |
128.64 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11448396
|
Thermo Scientific Alfa Aesar
A15415.0B |
1000 g | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
6276-54-6 | |
130.012 | |
IHPRVZKJZGXTBQ-UHFFFAOYSA-N | |
11469095 | |
C(CN)CCl.Cl |
C3H9Cl2N | |
MFCD00012913 | |
3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b | |
3-chloropropan-1-amine;hydrochloride |
Specifications
6276-54-6 | |
C3H9Cl2N | |
3667451 | |
IHPRVZKJZGXTBQ-UHFFFAOYSA-N | |
3-chloropropan-1-amine;hydrochloride | |
11469095 | |
98% |
147°C to 152°C | |
MFCD00012913 | |
3-chloropropylamine hydrochloride, 3-chloropropan-1-amine hydrochloride, 3-chloropropylamine hcl, 1-amino-3-chloropropane hydrochloride, 3-chloro-1-propanamine hydrochloride, 1-propanamine, 3-chloro-, hydrochloride, 3-chloropropylammonium chloride, acmc-1b7ws, chloropropylamine hydrochloride, ksc491e7b | |
C(CN)CCl.Cl | |
130.012 | |
130.02 | |
3-Chloropropylamine hydrochloride |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'einecsNumber' : 228-474-6
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
- 3-Chloropropylamine hydrochloride
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
- MIXTURE LIST-Contains : 3-Chloropropylaminehydrochloride
RUO – Research Use Only