missing translation for 'onlineSavingsMsg'
Learn More
3-Chloropyridine, 99%, Thermo Scientific Chemicals Product Code.: 10162230

3-Chloropyridine, 99%, Thermo Scientific Chemicals

Product Code. 10162230
25 mL, Glass bottle
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10162230

Brand: Thermo Scientific Acros 110020250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-60-8
Molecular Formula C5H4ClN
Molecular Weight (g/mol) 113.54
MDL Number MFCD00006375
InChI Key PWRBCZZQRRPXAB-UHFFFAOYSA-N
Synonym pyridine, 3-chloro, m-chloropyridine, unii-1m13huc1p4, ccris 1715, chloropyridine, b-chloropyridine, 3-chloro pyridine, 3-chloro-pyridine, 3-chloropyridine, 3-chloranylpyridine
PubChem CID 12287
IUPAC Name 3-chloropyridine
SMILES ClC1=CC=CN=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.1900g/mL
Boiling Point 147.0°C to 150.0°C
Flash Point 66°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C5H4N)Cl
Refractive Index 1.5310 to 1.5340
Quantity 25 mL
Beilstein 20, 230
Specific Gravity 1.19
Solubility Information Solubility in water: 10g/L (20°C)
Formula Weight 113.55
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 3-Chloropyridine
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0465074 Certificate TypeCertificate of Analysis Date13/08/2024 Catalog Number110020250,110021000
Lot NumberA0456090 Certificate TypeCertificate of Analysis Date07/09/2023 Catalog Number110020250,110021000
Lot NumberA0452028 Certificate TypeCertificate of Analysis Date02/05/2023 Catalog Number110020250,110021000
Lot NumberA0437105 Certificate TypeCertificate of Analysis Date23/12/2021 Catalog Number110020250,110021000
Lot NumberA0432971 Certificate TypeCertificate of Analysis Date16/08/2021 Catalog Number110020250,110021000
compliance-icons
Product Identifier
  • 3-Chloropyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloropyridine, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.