missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophene-2-carbonyl chloride, Thermo Scientific™

Product Code. 10039261
Click to view available options
Quantity:
250 mg
1 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 86427-02-3
Molecular Formula C5H2Cl2OS
Molecular Weight (g/mol) 181.03
MDL Number MFCD00068137
InChI Key GCPHKTQMABHWPY-UHFFFAOYSA-N
Synonym 3-chlorothiophene-2-carbonylchloride, 3-chloro-thiophene-2-carbonyl chloride, 3-chloro-2-chlorocarbonyl thiophene, 2-thiophenecarbonyl chloride, 3-chloro-9ci, pubchem5480, timtec-bb sbb005473, buttpark 29\06-44, chloro thiophene-2-carbonyl chloride, 3-chlorothiophene-2-carboyl chloride, 3-chloro-2-thiophenecarbonyl chloride
PubChem CID 2735824
IUPAC Name 3-chlorothiophene-2-carbonyl chloride
SMILES ClC(=O)C1=C(Cl)C=CS1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 250 mg
Formula Weight 181.04
Percent Purity 97%
Chemical Name or Material 3-Chlorothiophene-2-carbonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.