missing translation for 'onlineSavingsMsg'
Learn More

3-Chlorothiophene, 96%, Thermo Scientific™

Product Code. 10478612
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 17249-80-8
Molecular Formula C4H3ClS
Molecular Weight (g/mol) 118.59
MDL Number MFCD00043887
InChI Key QUBJDMPBDURTJT-UHFFFAOYSA-N
Synonym thiophene, 3-chloro, 3-chloro thiophene, 3-thienyl chloride, 3-chlorthiophen, 3-chloro-thiophene, 3-chloranylthiophene, pubchem5477, acmc-1cdmv, 3-chlorothiophene, ksc175g2h
PubChem CID 87017
IUPAC Name 3-chlorothiophene
SMILES C1=CSC=C1Cl

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Density 1.2760g/mL
Boiling Point 137.0°C to 139.0°C
Flash Point 37°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Refractive Index 1.5520 to 1.5540
Quantity 25 g
Specific Gravity 1.276
Solubility Information Solubility in water: <0.2g/L (20°C).
Formula Weight 118.59
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Chlorothiophene, 98%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'