missing translation for 'onlineSavingsMsg'
Learn More

3-Cyano-2-methoxypyridine, 97+%, Thermo Scientific™

Product Code. 15429067
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Cyano-2-methoxypyridine is used as an organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7254-34-4
Molecular Formula C7H6N2O
Molecular Weight (g/mol) 134.14
MDL Number MFCD07363804
InChI Key IHWZCGMZILLKFD-UHFFFAOYSA-N
PubChem CID 252099
IUPAC Name 2-methoxypyridine-3-carbonitrile
SMILES COC1=NC=CC=C1C#N

Specifications

Quantity 250 mg
Synonym 2-methoxynicotinonitrile, 3-cyano-2-methoxypyridine, 2-methoxy-nicotinonitrile, 2-methoxy-3-cyanopyridine, 3-pyridinecarbonitrile, 2-methoxy, 2-methoxy-3-pyridinecarbonitrile, 2-methoxynicotinonitril, acmc-1bcy7, 3-pyridinecarbonitrile,2-methoxy
Solubility Information Slightly soluble in water.
Formula Weight 134.14
Percent Purity ≥97%
Chemical Name or Material 3-Cyano-2-methoxypyridine
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'