missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanobenzylsulfonyl chloride, 97%, Thermo Scientific™

Product Code. 10153364
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 56106-01-5
Molecular Formula C8H6ClNO2S
Molecular Weight (g/mol) 215.651
MDL Number MFCD09046692
InChI Key YNWYDZBBHOWDAE-UHFFFAOYSA-N
Synonym 3-cyanophenyl methanesulfonyl chloride, 3-cyanobenzylsulfonyl chloride, 3-cyanophenyl methanesulphonyl chloride, 3-chlorosulphonyl methyl benzonitrile, 3-cyano-phenyl-methanesulfonyl chloride, 3-cyanobenzylsulphonyl chloride, 3-cyano benzylsulfonyl chloride, benzenemethanesulfonylchloride, 3-cyano, 3-cyano-phenyl methanesulfonyl chloride, 3-chlorosulfonyl methyl benzenecarbonitrile
PubChem CID 16774314
IUPAC Name (3-cyanophenyl)methanesulfonyl chloride
SMILES C1=CC(=CC(=C1)CS(=O)(=O)Cl)C#N

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 215.66
Percent Purity 97%
Chemical Name or Material 3-Cyanobenzylsulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.