missing translation for 'onlineSavingsMsg'
Learn More

3-Cyanophenylboronic acid pinacol ester, 97%, Thermo Scientific Chemicals

Product Code. 10535870
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 214360-46-0
Molecular Formula C13H16BNO2
Molecular Weight (g/mol) 229.09
MDL Number MFCD03789262
InChI Key FIGQEPXOSAFKTA-UHFFFAOYSA-N
Synonym 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, 3-cyanophenylboronic acid pinacol ester, 3-cyanophenylboronic acid, pinacol ester, benzonitrile, 3-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl, 3-cyanobenzeneboronic acid pinacol ester, 3-tetramethyl-1,3,2-dioxaborolan-2-yl benzonitrile, pubchem15603, 3-cyanophenylboronicacidpinacolester, 3-cyanobenzeneboronic acid, pinacol ester
PubChem CID 2760590
IUPAC Name 3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile
SMILES CC1(C)OB(OC1(C)C)C1=CC=CC(=C1)C#N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 80.0°C to 81.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Formula Weight 229.09
Percent Purity 97%
Physical Form Crystalline Powder
Chemical Name or Material 3-Cyanophenylboronic acid pinacol ester
Show More Show Less
compliance-icons
Product Identifier
  • 3-Cyanophenylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'