missing translation for 'onlineSavingsMsg'
Learn More

3-Cyclopentylpropionyl chloride, 98%, Thermo Scientific Chemicals

64.02 € - 237.18 €

Chemical Identifiers

CAS 104-97-2
Molecular Formula C8H13ClO
Molecular Weight (g/mol) 160.64
MDL Number MFCD00001391
InChI Key SZQVEGOXJYTLLB-UHFFFAOYSA-N
Synonym 3-cyclopentylpropionyl chloride, cyclopentylpropionyl chloride, cyclopentanepropionyl chloride, 3-cyclopentyl propionyl chloride, cyclopentanepropanoyl-chloride, pubchem21363, cyclopentylpropionylchloride, cyclopentanepropanoylchloride, 3-cyclopentylpropanyl chloride, cyclopentane propionyl chloride
PubChem CID 66039
IUPAC Name 3-cyclopentylpropanoyl chloride
SMILES ClC(=O)CCC1CCCC1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11493298
View Documents
Thermo Scientific Alfa Aesar
A17715.14
25 g
64.02 €
25g
Please to purchase this item. Need a web account? Register with us today!
11403308
View Documents
Thermo Scientific Alfa Aesar
A17715.22
100 g
237.18 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

104-97-2
160.64
SZQVEGOXJYTLLB-UHFFFAOYSA-N
66039
ClC(=O)CCC1CCCC1
C8H13ClO
MFCD00001391
3-cyclopentylpropionyl chloride, cyclopentylpropionyl chloride, cyclopentanepropionyl chloride, 3-cyclopentyl propionyl chloride, cyclopentanepropanoyl-chloride, pubchem21363, cyclopentylpropionylchloride, cyclopentanepropanoylchloride, 3-cyclopentylpropanyl chloride, cyclopentane propionyl chloride
3-cyclopentylpropanoyl chloride

Specifications

104-97-2
199°C to 200°C
Acrid
1.4645
UN3265
Moisture sensitive
SZQVEGOXJYTLLB-UHFFFAOYSA-N
3-cyclopentylpropanoyl chloride
66039
98%
1.049
84°C (184°F)
C8H13ClO
MFCD00001391
1856952
3-cyclopentylpropionyl chloride, cyclopentylpropionyl chloride, cyclopentanepropionyl chloride, 3-cyclopentyl propionyl chloride, cyclopentanepropanoyl-chloride, pubchem21363, cyclopentylpropionylchloride, cyclopentanepropanoylchloride, 3-cyclopentylpropanyl chloride, cyclopentane propionyl chloride
ClC(=O)CCC1CCCC1
160.64
160.64
3-Cyclopentylpropionyl chloride
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.

P210-P235-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c

H227-H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'einecsNumber' : 203-257-9

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.