missing translation for 'onlineSavingsMsg'
Learn More

3-Decyne, 98+%, Thermo Scientific™

Product Code. 11409766
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2384-85-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00039987
InChI Key JUWXVJKQNKKRLD-UHFFFAOYSA-N
Synonym 3-decyne, ethyl n-hexyl acetylene, acmc-1cb3a
PubChem CID 75425
IUPAC Name dec-3-yne
SMILES CCCCCCC#CCC

Specifications

Density 0.767
Boiling Point 175°C to 176°C
Flash Point 56°C (132°F)
Odor Petrol-like
Refractive Index 1.435
Quantity 1 g
UN Number UN3295
Beilstein 1698698
Formula Weight 138.26
Percent Purity ≥98%
Chemical Name or Material 3-Decyne
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'