missing translation for 'onlineSavingsMsg'
Learn More

3-Decyne, 98+%, Thermo Scientific™

Product Code. p-7024985
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11409766

Brand: Thermo Scientific Alfa Aesar B20268.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2384-85-2
Molecular Formula C10H18
Molecular Weight (g/mol) 138.25
MDL Number MFCD00039987
InChI Key JUWXVJKQNKKRLD-UHFFFAOYSA-N
Synonym 3-decyne, ethyl n-hexyl acetylene, acmc-1cb3a
PubChem CID 75425
IUPAC Name dec-3-yne
SMILES CCCCCCC#CCC

Specifications

Density 0.767
Boiling Point 175°C to 176°C
Flash Point 56°C (132°F)
Odor Petrol-like
Refractive Index 1.435
Quantity 1 g
UN Number UN3295
Beilstein 1698698
Formula Weight 138.26
Percent Purity ≥98%
Chemical Name or Material 3-Decyne
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.