missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylamino-1,2,4-dithiazole-5-thione, 97%, Thermo Scientific™

30.30 € - 87.33 €

Chemical Identifiers

CAS 29220-04-0
Molecular Formula C4H6N2S3
Molecular Weight (g/mol) 178.286
MDL Number MFCD00052567
InChI Key IDRNGWAVAQNFSE-UHFFFAOYSA-N
Synonym 3-dimethylamino-1,2,4-dithiazole-5-thione, 5-dimethylamino-3h-1,2,4-dithiazole-3-thione, 5-dimethylamino-1,2,4-dithiazole-3-thione, bio9c6, acmc-20an52, 5-dimethylamino-1,2,4-dithiazoline-3-thione, 3h-1,2,4-dithiazole-3-thione, dimethylamino
PubChem CID 2747561
IUPAC Name 5-(dimethylamino)-1,2,4-dithiazole-3-thione
SMILES CN(C)C1=NC(=S)SS1
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11325319
View Documents
Thermo Scientific Alfa Aesar
B20574.03
1 g
30.30 €
1g
Please to purchase this item. Need a web account? Register with us today!
11335319
View Documents
Thermo Scientific Alfa Aesar
B20574.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
3-Dimethylamino-1,2,4-dithiazole-5-thione, is used as a pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Specifications

Chemical Identifiers

29220-04-0
178.286
IDRNGWAVAQNFSE-UHFFFAOYSA-N
2747561
CN(C)C1=NC(=S)SS1
C4H6N2S3
MFCD00052567
3-dimethylamino-1,2,4-dithiazole-5-thione, 5-dimethylamino-3h-1,2,4-dithiazole-3-thione, 5-dimethylamino-1,2,4-dithiazole-3-thione, bio9c6, acmc-20an52, 5-dimethylamino-1,2,4-dithiazoline-3-thione, 3h-1,2,4-dithiazole-3-thione, dimethylamino
5-(dimethylamino)-1,2,4-dithiazole-3-thione

Specifications

29220-04-0
C4H6N2S3
3-dimethylamino-1,2,4-dithiazole-5-thione, 5-dimethylamino-3h-1,2,4-dithiazole-3-thione, 5-dimethylamino-1,2,4-dithiazole-3-thione, bio9c6, acmc-20an52, 5-dimethylamino-1,2,4-dithiazoline-3-thione, 3h-1,2,4-dithiazole-3-thione, dimethylamino
IDRNGWAVAQNFSE-UHFFFAOYSA-N
5-(dimethylamino)-1,2,4-dithiazole-3-thione
2747561
97%
175°C to 177°C
MFCD00052567
Slightly soluble in water.
CN(C)C1=NC(=S)SS1
178.286
178.31
3-Dimethylamino-1,2,4-dithiazole-5-thione
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.