missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminobenzoic anhydride, 97%, Thermo Scientific™

53.97 € - 265.00 €

Chemical Identifiers

CAS 4629-50-9
Molecular Formula C18H20N2O3
Molecular Weight (g/mol) 312.37
MDL Number MFCD09757555
InChI Key ROXBMXDDDMXDFQ-UHFFFAOYSA-N
Synonym 3-dimethylaminobenzoic anhydride, 3-dimethylamino benzoyl 3-dimethylamino benzoate, 3-dimethylamino benzoic anhydride, bis 3-dimethylamino benzoic acid anhydride, 3-dimethylaminobenzoyl 3-dimethylaminobenzoate, benzoic acid,3-dimethylamino-, anhydride with 3-dimethylamino benzoic acid
PubChem CID 46779096
IUPAC Name [3-(dimethylamino)benzoyl] 3-(dimethylamino)benzoate
SMILES CN(C)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)N(C)C
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15495157
View Documents
Thermo Scientific Alfa Aesar
H32446.03
1 g
53.97 €
1g
Please to purchase this item. Need a web account? Register with us today!
15405167
View Documents
Thermo Scientific Alfa Aesar
H32446.09
10 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

4629-50-9
312.37
ROXBMXDDDMXDFQ-UHFFFAOYSA-N
46779096
CN(C)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)N(C)C
C18H20N2O3
MFCD09757555
3-dimethylaminobenzoic anhydride, 3-dimethylamino benzoyl 3-dimethylamino benzoate, 3-dimethylamino benzoic anhydride, bis 3-dimethylamino benzoic acid anhydride, 3-dimethylaminobenzoyl 3-dimethylaminobenzoate, benzoic acid,3-dimethylamino-, anhydride with 3-dimethylamino benzoic acid
[3-(dimethylamino)benzoyl] 3-(dimethylamino)benzoate

Specifications

4629-50-9
C18H20N2O3
3-dimethylaminobenzoic anhydride, 3-dimethylamino benzoyl 3-dimethylamino benzoate, 3-dimethylamino benzoic anhydride, bis 3-dimethylamino benzoic acid anhydride, 3-dimethylaminobenzoyl 3-dimethylaminobenzoate, benzoic acid,3-dimethylamino-, anhydride with 3-dimethylamino benzoic acid
CN(C)C1=CC=CC(=C1)C(=O)OC(=O)C1=CC(=CC=C1)N(C)C
312.37
312.37
3-Dimethylaminobenzoic anhydride
110°C to 114°C
MFCD09757555
ROXBMXDDDMXDFQ-UHFFFAOYSA-N
[3-(dimethylamino)benzoyl] 3-(dimethylamino)benzoate
46779096
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.