missing translation for 'onlineSavingsMsg'
Learn More

3-Dimethylaminophenol, 97+%, Thermo Scientific Chemicals

Product Code. 11486256
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 99-07-0
Molecular Formula C8H11NO
Molecular Weight (g/mol) 137.18
MDL Number MFCD00002264
InChI Key MESJRHHDBDCQTH-UHFFFAOYSA-N
Synonym 3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol
PubChem CID 7421
IUPAC Name 3-(dimethylamino)phenol
SMILES CN(C)C1=CC=CC(O)=C1

Specifications

Melting Point 80°C to 84°C
Boiling Point 267°C
Flash Point 148°C (298°F)
Odor Pungent
Quantity 25 g
Beilstein 774631
Sensitivity Light sensitive
Solubility Information Slightly soluble in water.
Formula Weight 137.18
Percent Purity ≥97%
Chemical Name or Material 3-Dimethylaminophenol
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'