Learn More
3-Dimethylaminophenol, 97+%, Thermo Scientific Chemicals
26.10 € - 251.00 €
Chemical Identifiers
CAS | 99-07-0 |
---|---|
Molecular Formula | C8H11NO |
Molecular Weight (g/mol) | 137.18 |
MDL Number | MFCD00002264 |
InChI Key | MESJRHHDBDCQTH-UHFFFAOYSA-N |
Synonym | 3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol |
PubChem CID | 7421 |
IUPAC Name | 3-(dimethylamino)phenol |
SMILES | CN(C)C1=CC=CC(O)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11476256
|
Thermo Scientific Alfa Aesar
B23067.06 |
5 g |
26.10 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11486256
|
Thermo Scientific Alfa Aesar
B23067.14 |
25 g |
74.90 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11496256
|
Thermo Scientific Alfa Aesar
B23067.22 |
100 g |
251.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Dimethylaminophenol can be used as neostigmine bromide intermediate and dye intermediate.
Solubility
Slightly soluble in water.
Notes
Light sensitive. Store away from oxidizing agents, light.
Chemical Identifiers
99-07-0 | |
137.18 | |
MESJRHHDBDCQTH-UHFFFAOYSA-N | |
7421 | |
CN(C)C1=CC=CC(O)=C1 |
C8H11NO | |
MFCD00002264 | |
3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol | |
3-(dimethylamino)phenol |
Specifications
99-07-0 | |
267°C | |
Pungent | |
MFCD00002264 | |
Light sensitive | |
Slightly soluble in water. | |
CN(C)C1=CC=CC(O)=C1 | |
137.18 | |
137.18 | |
3-Dimethylaminophenol |
80°C to 84°C | |
148°C (298°F) | |
C8H11NO | |
774631 | |
3-dimethylamino phenol, phenol, 3-dimethylamino, m-dimethylamino phenol, m-dimethylaminophenol, n,n-dimethyl-m-aminophenol, n,n-dimethyl-3-aminophenol, phenol, m-dimethylamino, 3-hydroxyphenyl dimethylamine, 3-hydroxy-n,n-dimethylaniline, 3-n,n-dimethylaminophenol | |
MESJRHHDBDCQTH-UHFFFAOYSA-N | |
3-(dimethylamino)phenol | |
7421 | |
≥97% |
Safety and Handling
GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302+H312+H332-H315-H319-H335
missing translation for 'einecsNumber' : 202-727-0
missing translation for 'rtecsNumber' : SL1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only