missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-2-cyclohexen-1-one, 99%, Thermo Scientific Chemicals

Product Code. 11402337
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
This item is not returnable. View return policy

3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Ethoxy-2-cyclohexen-1-one is a useful intermediate for the preparation of 3-alkyl- or aryl-2-cyclohexen-1-ones, by addition of organometallic reagents and subsequent hydrolysis. It helps in kinetic enolate formation (LDA) and alkylation occurs at the 6-position, the Stork-Danheiser kinetic alkylation procedure, which has found extensive use in alicyclic synthesis.

Solubility
Slightly soluble in water.

Notes
Store at 4°C. Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5323-87-5
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
MDL Number MFCD00001580
InChI Key JWCFJPLIRVYENQ-UHFFFAOYSA-N
Synonym 3-ethoxy-2-cyclohexen-1-one, 3-ethoxycyclohex-2-enone, 2-cyclohexen-1-one, 3-ethoxy, 3-ethoxy-2-cyclohexenone, 3-ethoxy-2-cyclohexene-1-one, 3-ethoxycyclohex-2-ene-1-one, 1-ethoxycyclohexene-3-one, acmc-209l4y, 3-ethoxy-2-cyclohexen-one, 3-ethoxy-cyclohex-2-enone
PubChem CID 79216
IUPAC Name 3-ethoxycyclohex-2-en-1-one
SMILES CCOC1=CC(=O)CCC1

Specifications

Density 1.04
Boiling Point 76°C to 78°C (1 mmHg)
Flash Point 107°C (224°F)
Refractive Index 1.506
Quantity 250 g
Beilstein 2042049
Solubility Information Slightly soluble in water.
Formula Weight 140.18
Percent Purity 99%
Chemical Name or Material 3-Ethoxy-2-cyclohexen-1-one

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.