missing translation for 'onlineSavingsMsg'
Learn More

3-Ethoxy-5-fluorobenzeneboronic acid, 95%, Thermo Scientific™

Product Code. 15439197
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850589-53-6
Molecular Formula C8H10BFO3
Molecular Weight (g/mol) 183.97
MDL Number MFCD07363779
InChI Key OVVBLFYHZAMJIK-UHFFFAOYSA-N
Synonym 3-ethoxy-5-fluorobenzeneboronic acid, 3-ethoxy-5-fluorophenyl boronic acid, 5-ethoxy-3-fluorophenylboronic acid, boronic acid,b-3-ethoxy-5-fluorophenyl, 3-ethoxy-5-fluorophenylboronicacid, acmc-209q3b, 3-ethoxy-5-fluoro-benzeneboronic acid, 3-ethoxy-5-fluoro-phenyl boronic acid, 3-ethoxy-5-fluorobenzeneboronicacid, 3-ethoxy-5-fluoranyl-phenyl boronic acid
PubChem CID 44558175
IUPAC Name (3-ethoxy-5-fluorophenyl)boronic acid
SMILES CCOC1=CC(F)=CC(=C1)B(O)O

Specifications

Melting Point 155°C to 158°C
Quantity 250 mg
Formula Weight 183.97
Percent Purity 95%
Chemical Name or Material 3-Ethoxy-5-fluorobenzeneboronic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'