missing translation for 'onlineSavingsMsg'
Learn More

3-(Ethoxycarbonyl)benzeneboronic acid, 97%, Thermo Scientific Chemicals

Product Code. 11342479
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4334-87-6
Molecular Formula C9H11BO4
Molecular Weight (g/mol) 193.99
MDL Number MFCD02179444
InChI Key REHVCPNQQBDOJJ-UHFFFAOYSA-N
Synonym 3-ethoxycarbonyl phenylboronic acid, 3-ethoxycarbonylphenyl boronic acid, 3-carbethoxyphenylboronic acid, m-ethoxycarbonylphenylboronic acid, 3-ethoxycarbonyl phenyl boronic acid, ethyl 3-dihydroxyboranyl benzoate, ethyl 3-boronobenzoate, 3-ethoxycarbonylbenzeneboronic acid, 3-ethoxycarbonyl benzeneboronic acid
PubChem CID 3249312
IUPAC Name (3-ethoxycarbonylphenyl)boronic acid
SMILES CCOC(=O)C1=CC=CC(=C1)B(O)O

Specifications

Melting Point 135°C to 139°C
Quantity 1 g
Formula Weight 193.99
Percent Purity 97%
Chemical Name or Material 3-(Ethoxycarbonyl)benzeneboronic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'