missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyl-3-oxetanemethanol, 96%, Thermo Scientific™

Product Code. 10130613
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 3047-32-3
Molecular Weight (g/mol) 116.16
MDL Number MFCD00190143
InChI Key UNMJLQGKEDTEKJ-UHFFFAOYSA-N
Synonym 3-ethyl-3-oxetanemethanol, 3-ethyloxetan-3-yl methanol, 3-oxetanemethanol, 3-ethyl, 3-ethyloxetane-3-methanol, unii-622325vbyo, 3-ethyl-3-hydroxymethyloxetane, 3-hydroxymethyl-3-ethyloxetane, 3-ethyl-3-hydroxymethyl oxetane, 3-ethyl-oxetan-3-yl-methanol, 2-hydroxymethyl-2-ethyl-1,3-epoxypropane
PubChem CID 76444
IUPAC Name (3-ethyloxetan-3-yl)methanol
SMILES CCC1(COC1)CO

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Colorless
Density 1.0190g/mL
Boiling Point 96.0°C (4.0 mmHg)
Flash Point 108°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass bottle
Molecular Formula C6H12O2
Refractive Index 1.4520 to 1.4540
Quantity 1 g
Specific Gravity 1.019
Formula Weight 116.16
Percent Purity 96%
Physical Form Liquid
Chemical Name or Material 3-Ethyl-3-oxetanemethanol
Show More Show Less
compliance-icons
Product Identifier
  • 3-Ethyl-3-oxetanemethanol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'