missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Specifications
Specifications
Boiling Point | 171°C |
Flash Point | 31°C (88°F) |
Quantity | 25 g |
UN Number | UN1224 |
Synonym | 3-ethylcyclopentanone, cyclopentanone, 3-ethyl, 3-ethyl cyclopentanone, cyclopentanone,3-ethyl, acmc-20an48, 3-ethyl-cyclopentan-1-one |
Solubility Information | Soluble in water. |
Formula Weight | 112.17 |
Percent Purity | 99% |
Chemical Name or Material | 3-Ethylcyclopentanone |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals >
Spot an opportunity for improvement?Share a Content Correction