Learn More
3-Ethylcyclopentanone, 99%, Thermo Scientific Chemicals
54.97 € - 751.74 €
Chemical Identifiers
CAS | 10264-55-8 |
---|---|
Molecular Formula | C7H12O |
Molecular Weight (g/mol) | 112.172 |
MDL Number | MFCD00045445 |
InChI Key | XERALSLWOPMNRJ-UHFFFAOYSA-N |
PubChem CID | 139121 |
IUPAC Name | 3-ethylcyclopentan-1-one |
SMILES | CCC1CCC(=O)C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11404387
|
Thermo Scientific Alfa Aesar
B20432.03 |
1 g |
54.97 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11414387
|
Thermo Scientific Alfa Aesar
B20432.06 |
5 g |
212.06 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11424387
|
Thermo Scientific Alfa Aesar
B20432.14 |
25 g |
751.74 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Ethylcyclopentanone, is used as a chemical reagent, pharmaceutical reagent and pharmaceutical intermediate.
Solubility
Soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.
Chemical Identifiers
10264-55-8 | |
112.172 | |
XERALSLWOPMNRJ-UHFFFAOYSA-N | |
3-ethylcyclopentan-1-one |
C7H12O | |
MFCD00045445 | |
139121 | |
CCC1CCC(=O)C1 |
Specifications
10264-55-8 | |
31°C (88°F) | |
MFCD00045445 | |
3-ethylcyclopentanone, cyclopentanone, 3-ethyl, 3-ethyl cyclopentanone, cyclopentanone,3-ethyl, acmc-20an48, 3-ethyl-cyclopentan-1-one | |
XERALSLWOPMNRJ-UHFFFAOYSA-N | |
3-ethylcyclopentan-1-one | |
139121 | |
99% |
171°C | |
C7H12O | |
UN1224 | |
Soluble in water. | |
CCC1CCC(=O)C1 | |
112.172 | |
112.17 | |
3-Ethylcyclopentanone |
Safety and Handling
GHS H Statement
H226
Flammable liquid and vapour.
P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
H226
missing translation for 'dotInformation' : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.