missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Ethyltoluene, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.
Solubility
Not miscible or difficult to mix in water.
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
Specifications
Specifications
Melting Point | -96°C |
Density | 0.865 |
Boiling Point | 158°C to 159°C |
Flash Point | 38°C (100°F) |
Refractive Index | 1.496 |
Quantity | 5 g |
UN Number | UN3295 |
Beilstein | 1850821 |
Solubility Information | Not miscible or difficult to mix in water. |
Formula Weight | 120.2 |
Show More |
Safety and Handling
Product Identifier
- 3-Ethyltoluene
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H226-Flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P264-Wash face, hands and anyexposed skin thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P331-Do NOT induce vomiting.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- MIXTURE LIST-Contains : m-Ethyltoluene
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction